Assay Details
| Gene Symbol: CD300LF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027974035.1 | XP_027829836.1 | 1 - 2 | 183 | 81 | |
| XM_027974036.1 | XP_027829837.1 | 1 - 2 | 183 | 81 | ||
| XM_027974037.1 | XP_027829838.1 | 1 - 2 | 209 | 81 | ||
More Information
Set Membership: |
|

CD300LF
Genomic Map