Assay Details
| Gene Symbol: SNRK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027958069.1 | XP_027813870.1 | 6 - 7 | 943 | 63 | |
| XM_027958070.1 | XP_027813871.1 | 4 - 5 | 985 | 63 | ||
| XM_027958071.1 | XP_027813872.1 | 3 - 4 | 906 | 63 | ||
| XM_027958072.1 | XP_027813873.1 | 6 - 7 | 1080 | 63 | ||
| XM_027958073.1 | XP_027813874.1 | 6 - 7 | 1016 | 63 | ||
| XM_027958074.1 | XP_027813875.1 | 4 - 5 | 811 | 63 | ||
| XM_027958075.1 | XP_027813876.1 | 5 - 6 | 884 | 63 | ||
More Information
Set Membership: |
|

SNRK
Genomic Map