Assay Details
| Gene Symbol: LOC101121661 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012129529.3 | XP_011984919.2 | 10 - 11 | 1197 | 79 | |
| XM_012129535.2 | XP_011984925.2 | 10 - 11 | 992 | 79 | ||
More Information
Set Membership: |
|

LOC101121661
Genomic Map