Assay Details
| Gene Symbol: LOC101121661 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012129529.3 | XP_011984919.2 | 2 - 3 | 403 | 90 | |
| XM_012129535.2 | XP_011984925.2 | 1 - 2 | 83 | 90 | ||
| XM_027964634.1 | XP_027820435.1 | 2 - 3 | 407 | 90 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_012129535.2 transcripts.
Set Membership: |
|

LOC101121661
Genomic Map