Assay Details
| Gene Symbol: LOC101121661 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012129529.3 | XP_011984919.2 | 9 - 10 | 1132 | 97 | |
| XM_012129535.2 | XP_011984925.2 | 9 - 10 | 927 | 97 | ||
| XM_027964634.1 | XP_027820435.1 | 9 - 10 | 1136 | 97 | ||
More Information
Set Membership: |
|

LOC101121661
Genomic Map