Assay Details
| Gene Symbol: UFD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027956920.1 | XP_027812721.1 | 4 - 5 | 535 | 56 | |
| XM_027956921.1 | XP_027812722.1 | 4 - 5 | 512 | 56 | ||
| XM_027956922.1 | XP_027812723.1 | 4 - 5 | 653 | 56 | ||
| XM_027956923.1 | XP_027812724.1 | 4 - 5 | 561 | 56 | ||
| XM_027956924.1 | XP_027812725.1 | 4 - 5 | 653 | 56 | ||
| XM_027956925.1 | XP_027812726.1 | 4 - 5 | 587 | 56 | ||
More Information
Set Membership: |
|

UFD1
Genomic Map