Assay Details
| Gene Symbol: WDR97 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027973477.1 | XP_027829278.1 | 7 - 8 | 1994 | 93 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

WDR97| Gene Symbol: WDR97 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027973477.1 | XP_027829278.1 | 7 - 8 | 1994 | 93 | |