Assay Details
| Gene Symbol: LOC101120372 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027960336.1 | XP_027816137.1 | 1 - 2 | 46 | 122 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101120372| Gene Symbol: LOC101120372 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027960336.1 | XP_027816137.1 | 1 - 2 | 46 | 122 | |
Set Membership: |
|