Assay Details
| Gene Symbol: PHGDH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027973691.1 | XP_027829492.1 | 11 - 12 | 1547 | 55 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PHGDH| Gene Symbol: PHGDH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027973691.1 | XP_027829492.1 | 11 - 12 | 1547 | 55 | |
Set Membership: |
|