Assay Details
| Gene Symbol: NINJ1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004004069.4 | XP_004004118.1 | 1 - 2 | 390 | 99 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NINJ1| Gene Symbol: NINJ1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004004069.4 | XP_004004118.1 | 1 - 2 | 390 | 99 | |
Set Membership: |
|