Assay Details
| Gene Symbol: LIPT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004006144.4 | XP_004006193.4 | 1 - 2 | 70 | 96 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004006144.4 transcripts.
Set Membership: |
|

LIPT1
Genomic Map