Assay Details
| Gene Symbol: LOC101109268 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004008420.3 | XP_004008469.3 | 1 - 2 | 163 | 81 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101109268| Gene Symbol: LOC101109268 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004008420.3 | XP_004008469.3 | 1 - 2 | 163 | 81 | |