Assay Details
| Gene Symbol: IARS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012116190.2 | XP_011971580.1 | 20 - 21 | 2241 | 65 | |
| XM_012116202.2 | XP_011971592.1 | 20 - 21 | 2241 | 65 | ||
More Information
Set Membership: |
|

IARS1
Genomic Map