Assay Details
Gene Symbol: ZNF793 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012190325.2 | XP_012045715.2 | 3 - 4 | 255 | 80 | |
XM_015099980.2 | XP_014955466.2 | 5 - 6 | 540 | 80 | ||
XM_015099981.2 | XP_014955467.2 | 5 - 6 | 530 | 80 | ||
XM_027977431.1 | XP_027833232.1 | 4 - 5 | 1798 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
