Assay Details
| Gene Symbol: LOC114115347 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027971206.1 | XP_027827007.1 | 3 - 4 | 222 | 100 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_027971206.1 transcripts.
Set Membership: |
|

LOC114115347
Genomic Map