Assay Details
Gene Symbol: ZNF516 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027961048.1 | XP_027816849.1 | 4 - 5 | 3495 | 88 | |
XM_027961049.1 | XP_027816850.1 | 4 - 5 | 4443 | 88 | ||
XM_027961050.1 | XP_027816851.1 | 4 - 5 | 3420 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
