Assay Details
| Gene Symbol: SLC35A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004002227.4 | XP_004002276.2 | 4 - 5 | 2035 | 84 | |
| XM_012177086.2 | XP_012032476.1 | 4 - 5 | 1904 | 84 | ||
| XM_027972310.1 | XP_027828111.1 | 4 - 5 | 628 | 84 | ||
More Information
Set Membership: |
|

SLC35A3
Genomic Map