Assay Details
| Gene Symbol: CD69 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004006885.4 | XP_004006934.1 | 1 - 2 | 139 | 98 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: CD69 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004006885.4 | XP_004006934.1 | 1 - 2 | 139 | 98 | |