Assay Details
| Gene Symbol: SLC25A41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004008574.4 | XP_004008623.3 | 3 - 4 | 2875 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLC25A41| Gene Symbol: SLC25A41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004008574.4 | XP_004008623.3 | 3 - 4 | 2875 | 64 | |
Set Membership: |
|