Assay Details
| Gene Symbol: PLN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004011166.4 | XP_004011215.1 | 1 - 2 | 108 | 95 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004011166.4 transcripts.
Set Membership: |
|

PLN
Genomic Map