Assay Details
Gene Symbol: TTI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004014563.4 | XP_004014612.2 | 7 - 8 | 3191 | 69 | |
XM_012189178.2 | XP_012044568.2 | 8 - 9 | 3243 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
