Assay Details
| Gene Symbol: PGBD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012104935.3 | XP_011960325.2 | 6 - 7 | 1382 | 72 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PGBD5| Gene Symbol: PGBD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012104935.3 | XP_011960325.2 | 6 - 7 | 1382 | 72 | |
Set Membership: |
|