Assay Details
| Gene Symbol: LOC101109952 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012175114.3 | XP_012030504.2 | 4 - 5 | 427 | 94 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101109952| Gene Symbol: LOC101109952 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012175114.3 | XP_012030504.2 | 4 - 5 | 427 | 94 | |
Set Membership: |
|