Assay Details
| Gene Symbol: LOC101109952 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012175114.3 | XP_012030504.2 | 6 - 7 | 677 | 129 | |
| Gene Symbol: LOC114114041 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027967797.1 | XP_027823598.1 | 1 - 2 | 136 | 129 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_027967797.1 transcripts.
Set Membership: |
|

LOC101109952
Genomic Map