Assay Details
| Gene Symbol: LOC101103111 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027960952.1 | XP_027816753.1 | 2 - 3 | 362 | 73 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101103111| Gene Symbol: LOC101103111 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027960952.1 | XP_027816753.1 | 2 - 3 | 362 | 73 | |
Set Membership: |
|