Assay Details
Gene Symbol: LOC101103111 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027960952.1 | XP_027816753.1 | 6 - 7 | 864 | 70 |
Gene Symbol: LOC114110472 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027960951.1 | XP_027816752.1 | 3 - 4 | 324 | 70 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
