Assay Details
| Gene Symbol: WARS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027973655.1 | XP_027829456.1 | 4 - 5 | 1846 | 64 | |
| XM_027973662.1 | XP_027829463.1 | 4 - 5 | 1676 | 64 | ||
More Information
Set Membership: |
|

WARS2
Genomic Map