Assay Details
| Gene Symbol: LOC101105754 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027971104.1 | XP_027826905.1 | 2 - 3 | 419 | 71 | |
| XM_027971105.1 | XP_027826906.1 | 2 - 3 | 419 | 71 | ||
| XM_027971106.1 | XP_027826907.1 | 2 - 3 | 419 | 71 | ||
More Information
Set Membership: |
|

LOC101105754
Genomic Map