Assay Details
| Gene Symbol: LOC101105754 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027971104.1 | XP_027826905.1 | 5 - 6 | 908 | 75 | |
| XM_027971105.1 | XP_027826906.1 | 5 - 6 | 908 | 75 | ||
| XM_027971106.1 | XP_027826907.1 | 5 - 6 | 908 | 75 | ||

LOC101105754
Genomic Map