Assay Details
| Gene Symbol: DSE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012182542.3 | XP_012037932.1 | 4 - 5 | 1301 | 81 | |
| XM_015097337.2 | XP_014952823.1 | 3 - 4 | 563 | 81 | ||
| XM_027972351.1 | XP_027828152.1 | 4 - 5 | 1166 | 81 | ||
| XM_027972352.1 | XP_027828153.1 | 3 - 4 | 547 | 81 | ||
More Information
Set Membership: |
|

DSE
Genomic Map