Assay Details
| Gene Symbol: MMS22L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012182736.2 | XP_012038126.2 | 24 - 25 | 3893 | 70 | |
| XM_012182737.3 | XP_012038127.2 | 24 - 25 | 3897 | 70 | ||
More Information
Set Membership: |
|

MMS22L
Genomic Map