Assay Details
| Gene Symbol: LOC101106086 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027973838.1 | XP_027829639.1 | 9 - 10 | 1414 | 72 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101106086| Gene Symbol: LOC101106086 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027973838.1 | XP_027829639.1 | 9 - 10 | 1414 | 72 | |
Set Membership: |
|