Assay Details
| Gene Symbol: PRPF18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004014235.4 | XP_004014284.1 | 5 - 6 | 655 | 80 | |
| XM_012188400.3 | XP_012043790.1 | 6 - 7 | 682 | 80 | ||
| XM_012188402.2 | XP_012043792.1 | 6 - 7 | 540 | 80 | ||
More Information
Set Membership: |
|

PRPF18
Genomic Map