Assay Details
| Gene Symbol: LOC101111335 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004020949.4 | XP_004020998.2 | 15 - 16 | 2141 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101111335| Gene Symbol: LOC101111335 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004020949.4 | XP_004020998.2 | 15 - 16 | 2141 | 63 | |
Set Membership: |
|