Assay Details
| Gene Symbol: B3GALT6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004023657.4 | XP_004023706.3 | 1 | 739 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

B3GALT6| Gene Symbol: B3GALT6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004023657.4 | XP_004023706.3 | 1 | 739 | 71 | |
Set Membership: |
|