Assay Details
| Gene Symbol: CCNQ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012107098.2 | XP_011962488.1 | 6 - 7 | 853 | 59 | |
| XM_012107100.2 | XP_011962490.1 | 5 - 6 | 655 | 59 | ||
| XM_027962984.1 | XP_027818785.1 | 5 - 6 | 810 | 59 | ||
| XM_027962988.1 | XP_027818789.1 | 6 - 7 | 1464 | 59 | ||
More Information
Set Membership: |
|

CCNQ
Genomic Map