Assay Details
| Gene Symbol: ZNHIT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012185251.3 | XP_012040641.1 | 2 - 3 | 180 | 61 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNHIT3| Gene Symbol: ZNHIT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012185251.3 | XP_012040641.1 | 2 - 3 | 180 | 61 | |
Set Membership: |
|