Assay Details
| Gene Symbol: SEPTIN11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027971109.1 | XP_027826910.1 | 2 - 3 | 277 | 110 | |
| XM_027971110.1 | XP_027826911.1 | 2 - 3 | 277 | 110 | ||
| XM_027971111.1 | XP_027826912.1 | 2 - 3 | 277 | 110 | ||
| XM_027971113.1 | XP_027826914.1 | 2 - 3 | 277 | 110 | ||
| XM_027971114.1 | XP_027826915.1 | 2 - 3 | 277 | 110 | ||
| XM_027971115.1 | XP_027826916.1 | 2 - 3 | 277 | 110 | ||
| XM_027971116.1 | XP_027826917.1 | 2 - 3 | 277 | 110 | ||
| XM_027971117.1 | XP_027826918.1 | 2 - 3 | 277 | 110 | ||
| XM_027971118.1 | XP_027826919.1 | 2 - 3 | 277 | 110 | ||
| XM_027971119.1 | XP_027826920.1 | 2 - 3 | 277 | 110 | ||
| XM_027971120.1 | XP_027826921.1 | 2 - 3 | 277 | 110 | ||
More Information
Set Membership: |
|

SEPTIN11
Genomic Map