Assay Details
| Gene Symbol: C2CD6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027965092.1 | XP_027820893.1 | 3 - 4 | 375 | 98 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

C2CD6| Gene Symbol: C2CD6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027965092.1 | XP_027820893.1 | 3 - 4 | 375 | 98 | |