Assay Details
| Gene Symbol: C1H1orf43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004002552.4 | XP_004002601.2 | 2 - 3 | 419 | 106 | |
| XM_012182428.2 | XP_012037818.2 | 2 - 3 | 419 | 106 | ||
| XM_012182435.3 | XP_012037825.2 | 2 - 3 | 419 | 106 | ||
More Information
Set Membership: |
|

C1H1orf43
Genomic Map