Assay Details
| Gene Symbol: C1H1orf43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004002552.4 | XP_004002601.2 | 5 - 6 | 755 | 195 | |
| XM_004002553.4 | XP_004002602.2 | 4 - 5 | 640 | 195 | ||
| XM_012182428.2 | XP_012037818.2 | 5 - 6 | 755 | 282 | ||
| XM_012182435.3 | XP_012037825.2 | 5 - 6 | 755 | 229 | ||
| XM_012182438.2 | XP_012037828.2 | 4 - 5 | 650 | 229 | ||
More Information
Set Membership: |
|

C1H1orf43
Genomic Map