Assay Details
| Gene Symbol: LOC101108528 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004002634.4 | XP_004002683.1 | 3 - 4 | 425 | 77 | |
| XM_012183552.3 | XP_012038942.1 | 3 - 4 | 422 | 77 | ||
| XM_015092316.2 | XP_014947802.1 | 3 - 4 | 425 | 77 | ||
More Information
Set Membership: |
|

LOC101108528
Genomic Map