Assay Details
Gene Symbol: LOC101113728 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004004431.2 | XP_004004480.1 | 8 - 9 | 1534 | 57 | |
XM_012128319.2 | XP_011983709.1 | 9 - 10 | 1507 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
