Assay Details
| Gene Symbol: TIA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004005805.4 | XP_004005854.1 | 6 - 7 | 635 | 60 | |
| XM_004005806.4 | XP_004005855.1 | 5 - 6 | 602 | 60 | ||
| XM_012167439.3 | XP_012022829.1 | 6 - 7 | 642 | 60 | ||
| XM_012167453.3 | XP_012022843.1 | 5 - 6 | 609 | 60 | ||
| XM_012167463.3 | XP_012022853.1 | 6 - 7 | 648 | 60 | ||
| XM_027966652.1 | XP_027822453.1 | 6 - 7 | 652 | 60 | ||
| XM_027966653.1 | XP_027822454.1 | 6 - 7 | 655 | 60 | ||
| XM_027966654.1 | XP_027822455.1 | 6 - 7 | 645 | 60 | ||
| XM_027966655.1 | XP_027822456.1 | 5 - 6 | 619 | 60 | ||
| XM_027966656.1 | XP_027822457.1 | 5 - 6 | 612 | 60 | ||
| XM_027966657.1 | XP_027822458.1 | 6 - 7 | 645 | 60 | ||
| XM_027966658.1 | XP_027822459.1 | 6 - 7 | 638 | 60 | ||
| XM_027966659.1 | XP_027822460.1 | 5 - 6 | 604 | 60 | ||
More Information
Set Membership: |
|

TIA1
Genomic Map