Assay Details
| Gene Symbol: GPNMB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004007790.4 | XP_004007839.2 | 10 - 11 | 1689 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GPNMB| Gene Symbol: GPNMB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004007790.4 | XP_004007839.2 | 10 - 11 | 1689 | 71 | |
Set Membership: |
|