Assay Details
| Gene Symbol: RIC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004016178.4 | XP_004016227.2 | 5 - 6 | 671 | 84 | |
| XM_012095822.3 | XP_011951212.2 | 5 - 6 | 674 | 84 | ||
| XM_027979409.1 | XP_027835210.1 | 4 - 5 | 463 | 84 | ||
| XM_027979410.1 | XP_027835211.1 | 3 - 4 | 1759 | 84 | ||
More Information
Set Membership: |
|

RIC3
Genomic Map