Assay Details
| Gene Symbol: BUB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027960212.1 | XP_027816013.1 | 5 - 6 | 661 | 80 | |
| XM_027960213.1 | XP_027816014.1 | 5 - 6 | 661 | 80 | ||
More Information
Set Membership: |
|

BUB3
Genomic Map