Assay Details
| Gene Symbol: LOC101107495 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004020935.3 | XP_004020984.1 | 2 - 3 | 228 | 86 | |
| XM_004020936.3 | XP_004020985.1 | 2 - 3 | 210 | 86 | ||
| XM_012104412.2 | XP_011959802.2 | 5 - 6 | 951 | 86 | ||
| XM_012104415.2 | XP_011959805.1 | 2 - 3 | 483 | 86 | ||
More Information
Set Membership: |
|

LOC101107495
Genomic Map