Assay Details
| Gene Symbol: KRT9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004023155.4 | XP_004023204.2 | 1 - 2 | 475 | 97 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: KRT9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004023155.4 | XP_004023204.2 | 1 - 2 | 475 | 97 | |