Assay Details
| Gene Symbol: EPM2AIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027958009.1 | XP_027813810.1 | 1 | 1925 | 71 | |
| XM_027958010.1 | XP_027813811.1 | 1 | 1925 | 71 | ||
More Information
Set Membership: |
|

EPM2AIP1
Genomic Map